Learn More
Grainger Motor 1/6 2 HP 3 450/1 725 rpm 56Y 230V
Catalog No. 502765956
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 48 frame size and is thermally protected. This Century pool pump motor is designed for inground pools and spas and is made of corrosion resistant materials. It features permanently lubricated ball bearings and is CSA approved. The motor has a rotation speed of 3450 RPM.
Catalog No. 50-276-5956 Supplier Grainger Supplier No. 16U412
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 48 frame size and is thermally protected. This Century pool pump motor is designed for inground pools and spas and is made of corrosion resistant materials. It features permanently lubricated ball bearings and is CSA approved. The motor has a rotation speed of 3450 RPM.