Learn More
Grainger Motor 1 HP 3 450 rpm 56J 115/230V
Catalog No. 502765978
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement part for Century pool and spa pumps. It is made of thermoplastic and has a 1.5 HP, 3450 RPM motor. The motor face dimensions are 7-3/4" diameter x 5-1/2" long. It has a 1/2" diameter keyed shaft and is designed for 115V applications. The motor face attaches to the wet end housing and works with various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 1HP models. This is an OEM replacement part made to match the original pump manufacturer's specifications.
Catalog No. 50-276-5978 Supplier Grainger Supplier No. 16U477
The Century Pool and Spa Pump Motor Face is a replacement part for Century pool and spa pumps. It is made of thermoplastic and has a 1.5 HP, 3450 RPM motor. The motor face dimensions are 7-3/4" diameter x 5-1/2" long. It has a 1/2" diameter keyed shaft and is designed for 115V applications. The motor face attaches to the wet end housing and works with various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 1HP models. This is an OEM replacement part made to match the original pump manufacturer's specifications.