Learn More
Grainger Motor 2 HP 3 450 rpm 56C 115/230V
Catalog No. 502768288
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year manufacturer's warranty. It is designed for use in residential inground pools and spas and is CSA listed. The Century motor face should be installed by a qualified technician.
Catalog No. 50-276-8288 Supplier Grainger Supplier No. 16U472
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year manufacturer's warranty. It is designed for use in residential inground pools and spas and is CSA listed. The Century motor face should be installed by a qualified technician.