Learn More
Grainger Motor 2 HP 3 450 rpm 56J 208-230V
Catalog No. 502765393
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and has a 1/2 HP, 115/230 volt, 1725 RPM motor. It comes with a rear cover plate and is designed for inground pools and spas. The Century motor face provides a quality replacement part to get your pool or spa pump working again.
Catalog No. 50-276-5393 Supplier Grainger Supplier No. 16U396
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and has a 1/2 HP, 115/230 volt, 1725 RPM motor. It comes with a rear cover plate and is designed for inground pools and spas. The Century motor face provides a quality replacement part to get your pool or spa pump working again.