Learn More
Grainger Motor 2 HP 3 450 rpm 56Y 208-230V
Catalog No. 502765137
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures 7-1/2 inches in diameter and comes with a 1 year manufacturer's warranty. This Century motor face is designed for inground pools and spas and is easy to install.
Catalog No. 50-276-5137 Supplier Grainger Supplier No. 16U446
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B374, B43, B48L, B62, B73, 2HP and 3HP pumps. The motor face measures 7-1/2 inches in diameter and comes with a 1 year manufacturer's warranty. This Century motor face is designed for inground pools and spas and is easy to install.