Learn More
Grainger Motor 3/4 HP 3 450 rpm 48Y 208-230/460V
Catalog No. 502765765
Encompass_Preferred
The Century 16U462 is a 1/2 HP, 1725 RPM, 115/230V, single phase, open drip proof electric motor. It has a 48 frame size, CCW rotation (when viewed from shaft end), and features a cast iron body with a cast iron conduit box. The motor has Class F insulation and a 1.15 service factor. It can provide continuous duty operation and is thermally protected. The 16U462 is UL and CSA listed and meets NEMA MG-1 standards. With its versatile voltage and powerful performance, this motor is well-suited for a variety of industrial applications.
Catalog No. 50-276-5765 Supplier Grainger Supplier No. 16U462
The Century 16U462 is a 1/2 HP, 1725 RPM, 115/230V, single phase, open drip proof electric motor. It has a 48 frame size, CCW rotation (when viewed from shaft end), and features a cast iron body with a cast iron conduit box. The motor has Class F insulation and a 1.15 service factor. It can provide continuous duty operation and is thermally protected. The 16U462 is UL and CSA listed and meets NEMA MG-1 standards. With its versatile voltage and powerful performance, this motor is well-suited for a variety of industrial applications.