Learn More
Grainger Motor 3/4 HP 3 450 rpm 56CZ 115/230V
Catalog No. 502768351
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 56J frame size and is thermally protected. This Century pump motor is designed for inground pools and spas and is made with corrosion resistant construction. It has a 1.5" diameter shaft and is CSA approved. The motor operates at 3450 RPM.
Catalog No. 50-276-8351 Supplier Grainger Supplier No. 16U420
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a 56J frame size and is thermally protected. This Century pump motor is designed for inground pools and spas and is made with corrosion resistant construction. It has a 1.5" diameter shaft and is CSA approved. The motor operates at 3450 RPM.