Learn More
Grainger Motor 3/4 HP 3 450 rpm 56J 115/208-230V
Catalog No. 502766086
Encompass_Preferred
The Century Motor 3/4 HP 3,450 RPM 56J 115/208-230V is a single-phase, general purpose electric motor. It has a 3/4 horsepower rating and operates at 3450 RPM. The motor has a 56J frame size and can run on 115, 208, or 230 volt power. It is totally enclosed fan cooled (TEFC) with class F insulation and a 1.15 service factor. The motor is CSA certified and UL listed. It has a cast iron body, double sealed ball bearings, and a C-face mounting.
Catalog No. 50-276-6086 Supplier Grainger Supplier No. 16U399
The Century Motor 3/4 HP 3,450 RPM 56J 115/208-230V is a single-phase, general purpose electric motor. It has a 3/4 horsepower rating and operates at 3450 RPM. The motor has a 56J frame size and can run on 115, 208, or 230 volt power. It is totally enclosed fan cooled (TEFC) with class F insulation and a 1.15 service factor. The motor is CSA certified and UL listed. It has a cast iron body, double sealed ball bearings, and a C-face mounting.