Learn More
Grainger Motor 3/4 HP 3 450 rpm 56Y 115/230V
Catalog No. 502764986
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected. The motor is constructed with a corrosion-resistant stainless steel shaft and features permanently lubricated ball bearings for maintenance-free operation. With a 56Y frame, this rigid shaft motor is designed as a direct replacement for Hayward Super Pump and Super II pumps. It comes with a 1 year manufacturer's warranty.
Catalog No. 50-276-4986 Supplier Grainger Supplier No. 16U447
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected. The motor is constructed with a corrosion-resistant stainless steel shaft and features permanently lubricated ball bearings for maintenance-free operation. With a 56Y frame, this rigid shaft motor is designed as a direct replacement for Hayward Super Pump and Super II pumps. It comes with a 1 year manufacturer's warranty.