Learn More
Grainger Motor 3 HP 3 450 rpm 56C 230V
Catalog No. 502767700
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3 horsepower and 3450 RPM. The motor face comes with a one year limited warranty from Century. Overall, this is a quality replacement part for Century pool and spa pumps.
Catalog No. 50-276-7700 Supplier Grainger Supplier No. 16U473
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2 inches by 10-1/2 inches and has a 1/2 inch shaft diameter. It can handle up to 3 horsepower and 3450 RPM. The motor face comes with a one year limited warranty from Century. Overall, this is a quality replacement part for Century pool and spa pumps.