Learn More
Grainger Motor 3 HP 3 450 rpm 56J 208-230V
Catalog No. 502765915
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and attaches to the pump housing to complete the pump assembly. It features a 1/2 HP, 3450 RPM, 48Y frame motor that is totally enclosed and fan cooled (TEFC) for quiet operation and longevity. The motor face has a rear discharge configuration and includes a shaft and capacitor. It is designed for inground and aboveground pools and spas. This is a genuine OEM Century replacement part.
Catalog No. 50-276-5915 Supplier Grainger Supplier No. 16U401
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, N324, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and attaches to the pump housing to complete the pump assembly. It features a 1/2 HP, 3450 RPM, 48Y frame motor that is totally enclosed and fan cooled (TEFC) for quiet operation and longevity. The motor face has a rear discharge configuration and includes a shaft and capacitor. It is designed for inground and aboveground pools and spas. This is a genuine OEM Century replacement part.