Learn More
Grainger Motor 3 HP 3 450 rpm 56Y 208-230V
Catalog No. 502765301
Encompass_Preferred
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year limited warranty. It is designed for use in inground pools and spas and is easy to install. The Century motor face provides a quality replacement part to get your pool or spa pump working again.
Catalog No. 50-276-5301 Supplier Grainger Supplier No. 16U444
The Century Pool and Spa Pump Motor Face is a replacement motor face for Century pool and spa pumps. It is made of durable thermoplastic and fits various Century pump models including the B284, B232, B284F, B284S, and more. This motor face measures 10-1/2" diameter x 7-3/4" high and comes with a 1 year limited warranty. It is designed for use in inground pools and spas and is easy to install. The Century motor face provides a quality replacement part to get your pool or spa pump working again.