Learn More
Grainger Motor 4 HP 3 450 rpm 56Y 230V
Catalog No. 502768441
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1.5 HP, 3450 RPM pool pump motor designed for inground pools and spas. It features a 48Y frame, rigid base, and 1.5" diameter shaft. This single-phase motor has a voltage of 115/230 VAC and is totally enclosed and fan cooled. It can replace most major brand pool pump motors like Hayward, Pentair, and Sta-Rite. The motor dimensions are 9"L x 5.5"W x 7"H. It comes with a 1 year manufacturer's warranty. The Century pool pump motor provides reliable performance for circulating water in pools and spas.
Catalog No. 50-276-8441 Supplier Grainger Supplier No. 16U413
The Century Pool and Spa Pump Motor Rigid is a 1.5 HP, 3450 RPM pool pump motor designed for inground pools and spas. It features a 48Y frame, rigid base, and 1.5" diameter shaft. This single-phase motor has a voltage of 115/230 VAC and is totally enclosed and fan cooled. It can replace most major brand pool pump motors like Hayward, Pentair, and Sta-Rite. The motor dimensions are 9"L x 5.5"W x 7"H. It comes with a 1 year manufacturer's warranty. The Century pool pump motor provides reliable performance for circulating water in pools and spas.