Learn More
Grainger Motor 5 5/8 HP 3 450/1 725 rpm 56Y 230V
Catalog No. 502768613
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected for safety. The motor is totally enclosed and fan cooled (TEFC) with a 48 frame size. It has a 5/8" diameter shaft and measures approximately 9-1/2" in length. The motor is CSA listed for use with Century, Hayward, Pentair and Sta-Rite pool pumps.
Catalog No. 50-276-8613 Supplier Grainger Supplier No. 16U415
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected for safety. The motor is totally enclosed and fan cooled (TEFC) with a 48 frame size. It has a 5/8" diameter shaft and measures approximately 9-1/2" in length. The motor is CSA listed for use with Century, Hayward, Pentair and Sta-Rite pool pumps.