Learn More
Grainger Motor 5 HP 3 450 rpm 56Y 208-230V
Catalog No. 502765110
Encompass_Preferred
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected. The motor is totally enclosed and fan cooled (TEFC) with a 48 frame size. It has a 5/8" diameter shaft and measures 9-1/2" in overall length. The motor is CSA listed and comes with a 1 year limited warranty. It is designed for use with Century pool pumps.
Catalog No. 50-276-5110 Supplier Grainger Supplier No. 16U411
The Century Pool and Spa Pump Motor Rigid is a 1 HP, 115V, single phase pool pump motor with a rigid shaft. It has a rotation speed of 3450 RPM and is thermally protected. The motor is totally enclosed and fan cooled (TEFC) with a 48 frame size. It has a 5/8" diameter shaft and measures 9-1/2" in overall length. The motor is CSA listed and comes with a 1 year limited warranty. It is designed for use with Century pool pumps.