Learn More
Grainger Motor 5 HP 3 450 rpm 56Y 208-230/460V
Catalog No. 502765531
Encompass_Preferred
The Century Pool and Spa Pump Motor is a 1 HP, 115V, single phase pool pump motor. It has a 56Y frame, a 1.125" diameter shaft, and a CCW rotation. The motor has a service factor of 1.15 and operates at 3450 RPM. It is thermally protected and includes a capacitor. The motor is designed for inground pools and spas and is CSA approved. With its durable construction, this Century motor provides reliable performance for pumping applications.
Catalog No. 50-276-5531 Supplier Grainger Supplier No. 16U481
The Century Pool and Spa Pump Motor is a 1 HP, 115V, single phase pool pump motor. It has a 56Y frame, a 1.125" diameter shaft, and a CCW rotation. The motor has a service factor of 1.15 and operates at 3450 RPM. It is thermally protected and includes a capacitor. The motor is designed for inground pools and spas and is CSA approved. With its durable construction, this Century motor provides reliable performance for pumping applications.