Learn More
Ace Glass, Inc. Motor controller ac/dc input:120v 60Hz 10a max.
SDP
Catalog No. 50977997
Encompass
A compact, lightweight, solid state, ring stand mountable control unit, both AC and DC outlets. Rated 1 hp. for use with most stirrer motors. AC output: 120 volts, 10 amps. DC output: 0-120 volts, 6 amps.
Catalog No. 50-977-997 Supplier Ace Glass, Inc. Supplier No. 1353010
A compact, lightweight, solid state, ring stand mountable control unit, both AC and DC outlets. Rated 1 hp. for use with most stirrer motors. AC output: 120 volts, 10 amps. DC output: 0-120 volts, 6 amps.