Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Motor-Generator Model Set
GREENER_CHOICESCI_ED

Catalog No. S160138 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160138 3+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160138 Supplier Supertek™ Supplier No. PH50217
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Motor-Generator Set from Supertek™ helps students understand the connections between electric motors, generators, and transformers, both physically and technically.

  • This extensive modular system allows to set up different function models without tools:
    1. Various DC motors
    2. Series and shunt motor
    3. Synchronous motor
    4. DC generator and alternator
    5. Transformer
  • Allows the performance of 10 student experiments about the following topics:
    1. Magnetic field of a coil
    2. Conversion of electrical energy into kinetic energy
    3. Commutator
    4. Direct current motor
    5. Synchronous motor
    6. Series and shunt-wound motors
    7. Electromagnetic induction
    8. The electrical generator
    9. Engine-generator
    10. Transformer

Specifications

Grade 3+
Format Kit
Languages English
Subject Physics - Force, Motion and Energy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.