Learn More
Grainger Coxreels Motorized Hose Reel, Steel, 200 ft Capacity, 12V DC, 1 ea
Catalog No. 502765702
Encompass_Preferred
This motor-driven hose reel is engineered for durability and reliability in demanding industrial applications, including air, water, and oil lubrication transfer. Designed with a sturdy, one-piece all-welded steel frame, this unit ensures robust performance and long-term service life. Its motorized rewind system provides efficient, quiet, and smooth operation, while the low-profile riser and open drum slot facilitate non-crimping hose winding.
  • 1/3 hp 12V DC motor
  • 3,000 psi maximum operating pressure
  • -40°F to 250°F operating temperature range
  • Heavy-duty CNC robotically spun and ribbed steel discs
  • Permanently lubricated, self-aligning pillow block bearings
  • Adjustable tension brake for controlled retraction
  • Powder-coated, chip-resistant finish for corrosion protection
  • Versatile floor, vehicle, or wall-mount capability
Catalog No. 50-276-5702 Supplier Grainger Supplier No. 16X568
This motor-driven hose reel is engineered for durability and reliability in demanding industrial applications, including air, water, and oil lubrication transfer. Designed with a sturdy, one-piece all-welded steel frame, this unit ensures robust performance and long-term service life. Its motorized rewind system provides efficient, quiet, and smooth operation, while the low-profile riser and open drum slot facilitate non-crimping hose winding.
  • 1/3 hp 12V DC motor
  • 3,000 psi maximum operating pressure
  • -40°F to 250°F operating temperature range
  • Heavy-duty CNC robotically spun and ribbed steel discs
  • Permanently lubricated, self-aligning pillow block bearings
  • Adjustable tension brake for controlled retraction
  • Powder-coated, chip-resistant finish for corrosion protection
  • Versatile floor, vehicle, or wall-mount capability