Learn More
World Precision Instrument Motor Manipulator 3 Axes, ST-RT
SDP
Catalog No. 502539080
Encompass
Motorized Stereotaxic Manipulator Arms which are designed for fast, accurate positioning during stereotaxic surgery. Ideal for inexperienced users, the motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis. Manual movement available anytime. Optional Drill feature available.
Catalog No. 50-253-9080 Supplier World Precision Instrument Supplier No. 505378
Add to Cart
Add to Cart
Request a Quote
Motorized Stereotaxic Manipulator Arms which are designed for fast, accurate positioning during stereotaxic surgery. Ideal for inexperienced users, the motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis. Manual movement available anytime. Optional Drill feature available.