Learn More
Bellco Glass, Inc. Motor Maxon-Applkn 3L & 7L
SDP
Catalog No. 501946208
Encompass_Preferred
This allows a customer to use a bellco maxon motor/CellTrol ii controller with an Applikon vessel that normally uses p-100 motor. Motor mounting assembly with the following components Bellco motor assembly c542-a43a (1) Mounting plate: a505-62 (1) Mounting pegs: A505-622B (4) Flexible coupling: A505-612A (1), 4 screws. MADE IN THE USA
Catalog No. 50-194-6208 Supplier Bellco Glass, Inc. Supplier No. 776480037
This allows a customer to use a bellco maxon motor/CellTrol ii controller with an Applikon vessel that normally uses p-100 motor. Motor mounting assembly with the following components Bellco motor assembly c542-a43a (1) Mounting plate: a505-62 (1) Mounting pegs: A505-622B (4) Flexible coupling: A505-612A (1), 4 screws. MADE IN THE USA