Learn More
Caron Products And Services Motor, 115/208-230 VAC, 50/60 Hz, 1/15 Hp, 1 Amp, 1550/1300 RPM, 3/8" Diameter Shaft by 3 1/4" Long
Catalog No. NC2398213
Encompass_Preferred
Motor, 115/208-230 VAC, 50/60 Hz, 1/15 Hp, 1 Amp, 1550/1300 RPM, 3/8" Diameter Shaft by 3 1/4" Long
Catalog No. NC2398213 Supplier Caron Products And Services Supplier No. MTR130
Motor, 115/208-230 VAC, 50/60 Hz, 1/15 Hp, 1 Amp, 1550/1300 RPM, 3/8" Diameter Shaft by 3 1/4" Long