Learn More
Stoelting™ Motorized Lab Standard Stereotaxic
SDP
Catalog No. 10000001 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse, Neonatal Rat
Mouse, Rat
Rat
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
10-000-001 Rat
10-000-002 Mouse, Neonatal Rat
10-000-003 Mouse, Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-000-001 Supplier Stoelting™ Supplier No. 51600M
Add to Cart
Add to Cart
Request a Quote
Accuate positioning with a motorized manipulator arm and touchscreen display, easy-to-control movement with manual movement and optional drill feature available
  • Designed for fast, accurate positioning during stereotaxic surgery
  • Ideal for inexperienced users
  • Motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis
  • Manual movement available anytime
  • Automatic motorized movement using touchscreen display
  • Various speeds (3.0 mm/s to 0.15 μm/s*DV only)
  • 10 μm movement
Specifications
For Use With (Application) Stereotaxic Surgery
Animal Rat
Includes Base, u-frame, built-in warmer (controller extra), 3-axis motorized manipulator arm, touchscreen controller, corner-clamp probe holder, rat adaptor