Learn More
RWD LIFE SCIENCE INC MOTORIZED MICROMANIPULATOR
SDP
Catalog No. NC3404322
Encompass_Preferred
MM-500, an ultra-stable and compact motorized micromanipulator, is designed for probe positioning or pipettes holding with high precision, to perform in vivo and in vitro electrophysiology experiments, microinjection, etc.
Catalog No. NC3404322 Supplier RWD LIFE SCIENCE INC Supplier No. MM500R/L
Add to Cart
Add to Cart
Request a Quote
MM-500, an ultra-stable and compact motorized micromanipulator, is designed for probe positioning or pipettes holding with high precision, to perform in vivo and in vitro electrophysiology experiments, microinjection, etc.