Learn More
Chemglass Life Sciences Motorized Pipette Controller, Blue, AccuHelp
SDP
Catalog No. 5000980711
Encompass_Preferred
Speeds: High, Low and Gravity. Pipette Type: Glass or Plastic (0.1-100mL). Battery: Rechargeable. Battery Working Time: 10 hours continuous. Low Battery LED Indicator. Filter: Replaceable Membrane Filter. One Year Warranty. The rechargeable lightweight AccuHelp motorized pipettor combines advanced ergonomic design with precise liquid dispensing. The round shape, balanced weight, and light action spring buttons reduce fatigue and increase accuracy. AccuHelp is compatible with glass or plastic pipettes, ranging from 0.1 to 100mL and is available in four different colors. NOTE: There is a red light on the back of the pipette. If the red light turns on, that means the battery is low. There is also a red light on the charger itself. The red light on the charger means that the pipette is being charged. If the light on the charger turns green, the charging is complete.
Catalog No. 50-009-80711 Supplier Chemglass Life Sciences Supplier No. CLS3472E01
Speeds: High, Low and Gravity. Pipette Type: Glass or Plastic (0.1-100mL). Battery: Rechargeable. Battery Working Time: 10 hours continuous. Low Battery LED Indicator. Filter: Replaceable Membrane Filter. One Year Warranty. The rechargeable lightweight AccuHelp motorized pipettor combines advanced ergonomic design with precise liquid dispensing. The round shape, balanced weight, and light action spring buttons reduce fatigue and increase accuracy. AccuHelp is compatible with glass or plastic pipettes, ranging from 0.1 to 100mL and is available in four different colors. NOTE: There is a red light on the back of the pipette. If the red light turns on, that means the battery is low. There is also a red light on the charger itself. The red light on the charger means that the pipette is being charged. If the light on the charger turns green, the charging is complete.