Learn More
Scientific Industries TORBAL FTH100 Motorized Test Stand, with FB50 Force Gauge, Wireless Connectivity, 5 Application Modes, and over 15 Programmable Parameters
Catalog No. 502067582
Encompass_Preferred
Includes the TORBAL FB50 10lbF force gauge and all essential application modes: Peak, Cycle, Step, Advanced, and Manual. Fully automated and configurable for any tension or compression force testing application. Engineered with a reinforced column, an ultra-high-performance motor, as well as a heavy-duty cross-head to guarantee uninterrupted performance and continuous testing. Precise speed control from 0.4in to 11.8in per minute and 0.001in travel accuracy. The FTH has 11.8in of travel and a maximum load capacity of 100lbF. Configuring the test for your application is quick and easy with 200 testing profiles. Our intuitive dial knob control allows for an effortless navigation of menu options and input parameters. Included with the test stand: complete set of attachments, shaft extension, wireless antenna, mounting plate, grip knobs, and connection cables. Wireless connectivity between the test stand and force gauge means no tangled or loose cables.
Catalog No. 50-206-7582 Supplier Scientific Industries Supplier No. FTH100FB50
Includes the TORBAL FB50 10lbF force gauge and all essential application modes: Peak, Cycle, Step, Advanced, and Manual. Fully automated and configurable for any tension or compression force testing application. Engineered with a reinforced column, an ultra-high-performance motor, as well as a heavy-duty cross-head to guarantee uninterrupted performance and continuous testing. Precise speed control from 0.4in to 11.8in per minute and 0.001in travel accuracy. The FTH has 11.8in of travel and a maximum load capacity of 100lbF. Configuring the test for your application is quick and easy with 200 testing profiles. Our intuitive dial knob control allows for an effortless navigation of menu options and input parameters. Included with the test stand: complete set of attachments, shaft extension, wireless antenna, mounting plate, grip knobs, and connection cables. Wireless connectivity between the test stand and force gauge means no tangled or loose cables.