Learn More
Scientific Industries TORBAL FTV100 Motorized Test Stand, with FC10 Force Gauge, Wireless Connectivity, 5 Application Modes, and over 15 Programmable Parameters
Supplier:  Scientific Industries FTV100FC10
Encompass_Preferred
Includes the TORBAL FC10 2lbF force gauge and all essential application modes: Peak, Cycle, Step, Advanced, and Manual. Fully automated and configurable for any tension or compression force testing application. Engineered with a reinforced column, an ultra-high-performance motor, as well as a heavy-duty cross-head to guarantee uninterrupted performance and continuous testing. Precise speed control from 0.4in to 11.8in per minute and 0.001in travel accuracy. The FTV has 11.8in of travel and a maximum load capacity of 100lbF. Configuring the test for your application is quick and easy with 200 testing profiles. Our intuitive dial knob control allows for an effortless navigation of menu options and input parameters. Included with the test stand: complete set of attachments, shaft extension, wireless antenna, mounting plate, grip knobs, and connection cables. Wireless connectivity between the test stand and force gauge means no tangled or loose cables.
Catalog No. 50-206-7560