Learn More
Selleck Chemical LLC MOTS-c-5mg
SDP
Supplier:  Selleck Chemical LLC E20325MG
Encompass_Preferred
MOTS-c, a mitochondria-derived peptide (MDP), exerts antinociceptive and anti-inflammatory effects through activating AMPK pathway and inhibiting MAP kinases-c-fos signaling pathway.
Catalog No. 50-239-0024