Learn More
AdipoGen MOTS-c human
SDP
Catalog No. 502562433
Encompass_Preferred
Chemical. CAS 1627580-64-6. Formula C101H152N28O22S2 . C2HF3O2. MW 2174.6 . 114.0. Synthetic. Mitochondria-derived peptide MDP. Promotes biosynthesis of the endogenous AMP analog AICAR and consequently AMP-activated protein kinase AMPK. Induces cellular and systemic glucose uptake and improves insulin sensitivity. Shown to prevent diet-induced obesity DIO in mice. Potential anti-obesity and anti-aging compound.
Catalog No. 50-256-2433 Supplier AdipoGen Supplier No. AGCP30026M001
Chemical. CAS 1627580-64-6. Formula C101H152N28O22S2 . C2HF3O2. MW 2174.6 . 114.0. Synthetic. Mitochondria-derived peptide MDP. Promotes biosynthesis of the endogenous AMP analog AICAR and consequently AMP-activated protein kinase AMPK. Induces cellular and systemic glucose uptake and improves insulin sensitivity. Shown to prevent diet-induced obesity DIO in mice. Potential anti-obesity and anti-aging compound.