Learn More
Medchemexpress LLC MOTS-c (human) acetate | 99.7% | 2234.64 | 50 MG
SDP
Catalog No. 5000422914
Encompass_Preferred
MOTS-c (human) acetate is a blood-brain barrier-penetrating, mitochondrial-derived peptide that modulates the AMPK/PGC-1α pathway to enhance insulin sensitivity. It also regulates antioxidant and inflammatory pathways, promotes mitochondrial biogenesis, and improves glucose and lipid metabolism. It is studied for type 2 diabetes, traumatic brain injury, inflammatory diseases, and aging-related metabolic disorders.
  • Blood-brain barrier-penetrating mitochondrial peptide.
  • Enhances insulin sensitivity via AMPK/PGC-1α modulation.
  • Regulates antioxidant and inflammatory pathways.
  • Promotes mitochondrial biogenesis.
  • Improves glucose and lipid metabolism.
  • Anti-oxidative, anti-inflammatory, and neuroprotective.
  • Applications in diabetes, TBI, inflammatory, and aging studies.
Catalog No. 50-004-22914 Supplier Medchemexpress LLC Supplier No. HYP2048A50MG
MOTS-c (human) acetate is a blood-brain barrier-penetrating, mitochondrial-derived peptide that modulates the AMPK/PGC-1α pathway to enhance insulin sensitivity. It also regulates antioxidant and inflammatory pathways, promotes mitochondrial biogenesis, and improves glucose and lipid metabolism. It is studied for type 2 diabetes, traumatic brain injury, inflammatory diseases, and aging-related metabolic disorders.
  • Blood-brain barrier-penetrating mitochondrial peptide.
  • Enhances insulin sensitivity via AMPK/PGC-1α modulation.
  • Regulates antioxidant and inflammatory pathways.
  • Promotes mitochondrial biogenesis.
  • Improves glucose and lipid metabolism.
  • Anti-oxidative, anti-inflammatory, and neuroprotective.
  • Applications in diabetes, TBI, inflammatory, and aging studies.