Learn More
AdipoGen MOTS-c K14Q Mutant human
SDP
Catalog No. 502562423
Encompass_Preferred
Chemical. Formula C100H148N28O23S2 . C2HF3O2. MW 2174.6 . 114.0. Synthetic. The mitochondria-derived peptide MDP MOTS-c promotes biosynthesis of the endogenous AMP analog AICAR and consequently AMP-activated protein kinase AMPK. It induces cellular and systemic glucose uptake and improves insulin sensitivity and has been shown to prevent diet-induced obesity DIO in mice. This K14Q mutant of MOTS-c has been shown to be associated with exceptional longevity and may be interesting for aging research.
Catalog No. 50-256-2423 Supplier AdipoGen Supplier No. AGCP30033M001
Chemical. Formula C100H148N28O23S2 . C2HF3O2. MW 2174.6 . 114.0. Synthetic. The mitochondria-derived peptide MDP MOTS-c promotes biosynthesis of the endogenous AMP analog AICAR and consequently AMP-activated protein kinase AMPK. It induces cellular and systemic glucose uptake and improves insulin sensitivity and has been shown to prevent diet-induced obesity DIO in mice. This K14Q mutant of MOTS-c has been shown to be associated with exceptional longevity and may be interesting for aging research.