Learn More
Enterprise Technology Solutions Motsenbocker Advanced Adhesive Remover, f/Tape/Labels, Low-VOC, 32 oz, 6/CT, WE
SDP
Catalog No. 5000790343
Encompass_Preferred
Lift Off Tape, Label and Adhesive Remover can remove old and new tape, labels, adhesives, grease, oil gum and tar from a variety of surfaces, including carpets, upholstery, clothing and hard surfaces. Low-VOC, environmentally safe remover makes removal easier because it breaks the molecular bond between the adhesive and the surface. It works in conjunction with other Motsenbocker's Lift Off products.
Catalog No. 50-007-90343 Supplier Enterprise Technology Solutions Supplier No. MOT40703CT
Lift Off Tape, Label and Adhesive Remover can remove old and new tape, labels, adhesives, grease, oil gum and tar from a variety of surfaces, including carpets, upholstery, clothing and hard surfaces. Low-VOC, environmentally safe remover makes removal easier because it breaks the molecular bond between the adhesive and the surface. It works in conjunction with other Motsenbocker's Lift Off products.