Learn More
Enterprise Technology Solutions Motsenbocker Advanced Tape/Adhesive Remover,Spray,22 oz,Clear
SDP
Catalog No. 5000829812
Encompass_Preferred
Tape, Label and Adhesive Remover breaks the molecular bond for fast, easy removal of painter's tape, duct tape, mounting tape and weather stripping tape. Patented formula also removes spray adhesives, stickers, labels, chewing gum, sticky residues, wax and glue from hard surfaces. Spray around adhesive edges and wait at least 60 seconds. Use a scraper (not included) to agitate and lift the adhesive from the surface. Wipe clean to remove residue. Heavy-duty jobs may require more than one application. Low VOC formula uses less than half the regulated VOC limit for enhanced safety.
Catalog No. 50-008-29812 Supplier Enterprise Technology Solutions Supplier No. MOT40701
Tape, Label and Adhesive Remover breaks the molecular bond for fast, easy removal of painter's tape, duct tape, mounting tape and weather stripping tape. Patented formula also removes spray adhesives, stickers, labels, chewing gum, sticky residues, wax and glue from hard surfaces. Spray around adhesive edges and wait at least 60 seconds. Use a scraper (not included) to agitate and lift the adhesive from the surface. Wipe clean to remove residue. Heavy-duty jobs may require more than one application. Low VOC formula uses less than half the regulated VOC limit for enhanced safety.