Learn More
Andwin Scientific MOTTLER BRUSH 2.54 CM
SDP
Catalog No. NC9593428
Encompass_Preferred
The Mottler brush is ideal for dusting balances and weights. The brush is characterized as soft, thick goat hair brush and is non-scratching. It is held by a wide metal ferrule that is fastened onto a hardwood handle. This product has an overall length of 15.2 cm (6") and width of 2.5 cm (1").
Catalog No. NC9593428 Supplier Andwin Scientific Supplier No. 17230049
The Mottler brush is ideal for dusting balances and weights. The brush is characterized as soft, thick goat hair brush and is non-scratching. It is held by a wide metal ferrule that is fastened onto a hardwood handle. This product has an overall length of 15.2 cm (6") and width of 2.5 cm (1").