Learn More
ABBERIOR INSTRUMENTS AMERICA LLC MOUNT LIQUID ANTIFADE
Catalog No. NC1933858
Encompass_Preferred
abberior MOUNT LIQUID ANTIFADE is an aqueous mounting medium designed for 3D confocal and superresolution microscopy. The refractive index is identical to that of many biological samples (n = 1.38). abberior MOUNT LIQUID ANTIFADE is not hardening after embedding of specimen, to exclude changes of the 3D structure of samples during the hardening process.abberior MOUNT LIQUID ANTIFADE prevents photobleaching and preserves the signal of fluorescent labels. For scientific use only. Must not be used for diagnostic or medical purposes.
Catalog No. NC1933858 Supplier ABBERIOR INSTRUMENTS AMERICA LLC Supplier No. MM20092X15ML
abberior MOUNT LIQUID ANTIFADE is an aqueous mounting medium designed for 3D confocal and superresolution microscopy. The refractive index is identical to that of many biological samples (n = 1.38). abberior MOUNT LIQUID ANTIFADE is not hardening after embedding of specimen, to exclude changes of the 3D structure of samples during the hardening process.abberior MOUNT LIQUID ANTIFADE prevents photobleaching and preserves the signal of fluorescent labels. For scientific use only. Must not be used for diagnostic or medical purposes.