Learn More
Mitegen Small Aperture Assortment Beamline Sample Preparation Kit; 100 Dual Thickness MicroMounts (25 each of 10, 20, 30 and 50 um aperture DT MicroMounts) on standard 18 mm pins, Installed in 100 B1A-R bases, Packaged and stored in 5 MiTeGen base holders
SDP
Catalog No. 501938889
Encompass_Preferred
MiTeGen beamline sample kit; Contains 100 mounts, 25 pins of 4 different aperture sizes pre-installed in Reusable Goniometer bases
Catalog No. 50-193-8889 Supplier Mitegen Supplier No. BL100B1ARA1
MiTeGen beamline sample kit; Contains 100 mounts, 25 pins of 4 different aperture sizes pre-installed in Reusable Goniometer bases