Learn More
Mitegen Medium Aperture Assortment Beamline Sample Preparation Kit; 100 Dual Thickness MicroMounts (25 each of 75, 100, 150 and 200 um aperture DT MicroMounts) on standard 18 mm pins, Installed in 100 B5-R bases, Packaged and stored in 5 MiTeGen base holders
SDP
Catalog No. 501938893
Encompass_Preferred
MiTeGen beamline sample kit; Contains 100 mounts, 25 pins of 4 different aperture sizes pre-installed in Reusable Goniometer bases
Catalog No. 50-193-8893 Supplier Mitegen Supplier No. BL100B5RA2
MiTeGen beamline sample kit; Contains 100 mounts, 25 pins of 4 different aperture sizes pre-installed in Reusable Goniometer bases