Learn More
Glass Expansion Mount Nut for Guardian Probe SPS3/SPS4/AIMS
SDP
Catalog No. 502703037
Encompass_Preferred
Mount Nut for Guardian Probe for SPS3/SPS4/AIMS. The Mount Nut is compatible with the Guardian Probe for Aim Lab & Agilent SPS3/SPS4 Autosamplers, P/N 70-803-2008 (Probe Only) and 70-803-1957 (Prob with 0.75mm, Red, Probe Connecting Line).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3037 Supplier Glass Expansion Supplier No. 708031776
Mount Nut for Guardian Probe for SPS3/SPS4/AIMS. The Mount Nut is compatible with the Guardian Probe for Aim Lab & Agilent SPS3/SPS4 Autosamplers, P/N 70-803-2008 (Probe Only) and 70-803-1957 (Prob with 0.75mm, Red, Probe Connecting Line).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900