Learn More
Research Products International Corp Mount-Quick Mounting Media, 30 ml
SDP
Catalog No. 50550224
Encompass
Use Mount-Quick to preserve and protect laboratory specimens. Simple to use - apply thin coating to microscope slide and place coverslip over specimen. Mount-quick dries quickly and is ready for screening in minutes. Final preparation remains stable for a long period of time. Mount-Quick flows easily and eliminates trapping of air bubbles. Packaged in a convenient dispenser tube containing 30ml of media. Tube eliminates unnecessary transferring of media, only amount required is used. Mount-Quick can be removed with xylene if desired.
Catalog No. 50-550-224 Supplier Research Products International Corp Supplier No. 195700
Use Mount-Quick to preserve and protect laboratory specimens. Simple to use - apply thin coating to microscope slide and place coverslip over specimen. Mount-quick dries quickly and is ready for screening in minutes. Final preparation remains stable for a long period of time. Mount-Quick flows easily and eliminates trapping of air bubbles. Packaged in a convenient dispenser tube containing 30ml of media. Tube eliminates unnecessary transferring of media, only amount required is used. Mount-Quick can be removed with xylene if desired.