Learn More
Newcomer Supply Mount-Quick Mounting Media, 30cc tube. Excellent for tissue section transfer from one slide to another.
SDP
Catalog No. NC1168448
Encompass
Xylene Soluble Mounting Medium, 30cc tube Packaging allows maximum control during coverslipping. Fast drying. Clear. UNIQUE AS A TISSUE TRANSFER MEDIUM. Manufactured by Daido Sangyo Co. Ltd.
Catalog No. NC1168448 Supplier Newcomer Supply Supplier No. 6270
Xylene Soluble Mounting Medium, 30cc tube Packaging allows maximum control during coverslipping. Fast drying. Clear. UNIQUE AS A TISSUE TRANSFER MEDIUM. Manufactured by Daido Sangyo Co. Ltd.