Learn More
Newcomer Supply Mount-Quick Aqueous Mounting Media, 30cc
SDP
Catalog No. NC1835458
Encompass
Newcomer Supply Mount-Quick Aqueous is a ready-to-use liquid mounting medium for histological techniques requiring an aqueous based mounting medium solution and/or when dehydrating and clearing agents will adversely affect the stain, such as:  lipid/fat stains, crystal violet stains and solvent soluble immunoperoxidase chromogenic procedures. Mount-Quick Aqueous mounting medium is not recommended for use in immunofluorescence procedures.
Catalog No. NC1835458 Supplier Newcomer Supply Supplier No. 6271A
Newcomer Supply Mount-Quick Aqueous is a ready-to-use liquid mounting medium for histological techniques requiring an aqueous based mounting medium solution and/or when dehydrating and clearing agents will adversely affect the stain, such as:  lipid/fat stains, crystal violet stains and solvent soluble immunoperoxidase chromogenic procedures. Mount-Quick Aqueous mounting medium is not recommended for use in immunofluorescence procedures.