Learn More
Research Products International Corp Mount-Quick Aqueous Mounting Media, 30 ml
SDP
Catalog No. 50212451
Encompass
Use Mount-Quick to preserve and protect laboratory specimens. Simple to use - apply thin coating to microscope slide and place coverslip over specimen. Mount-quick dries quickly and is ready for screening in minutes. Final preparation remains stable for a long period of time. Mount-Quick flows easily and eliminates trapping of air bubbles. Packaged in a convenient dispenser tube containing 30ml of media. Tube eliminates unnecessary transferring of media, only amount required is used. Mount-Quick can be removed with xylene if desired. Mount-Quick Aqueous is a water-based mounting medium for use with liparoid/lipid sections, immunohistochemically stained sections and AEC method sections.
Catalog No. 50-212-451 Supplier Research Products International Corp Supplier No. 195705
Use Mount-Quick to preserve and protect laboratory specimens. Simple to use - apply thin coating to microscope slide and place coverslip over specimen. Mount-quick dries quickly and is ready for screening in minutes. Final preparation remains stable for a long period of time. Mount-Quick flows easily and eliminates trapping of air bubbles. Packaged in a convenient dispenser tube containing 30ml of media. Tube eliminates unnecessary transferring of media, only amount required is used. Mount-Quick can be removed with xylene if desired. Mount-Quick Aqueous is a water-based mounting medium for use with liparoid/lipid sections, immunohistochemically stained sections and AEC method sections.