Learn More
Electron Microscopy Sciences Mount Quick Water Base 30 ML
SDP
Catalog No. 5027424
Encompass_Preferred
Mounting media for cover glass.  It comes in a convenient dispensing tube or bottle.   Solvent based which is soluble in xylene and has a 10 minute dry time.  Its viscosity is 33 cps at 25 degree.
Catalog No. 50-274-24 Supplier Electron Microscopy Sciences Supplier No. 18002
Mounting media for cover glass.  It comes in a convenient dispensing tube or bottle.   Solvent based which is soluble in xylene and has a 10 minute dry time.  Its viscosity is 33 cps at 25 degree.