Learn More
Ted Pella Inc MOUNT SEM CYLINDER 50/PKMOUNT
SDP
Catalog No. NC9315727
Encompass
Specimen Mount, 12.2 x 10mm, JEOL, Aluminum, 100/PK
Catalog No. NC9315727 Supplier Ted Pella Inc Supplier No. 16231
Specimen Mount, 12.2 x 10mm, JEOL, Aluminum, 100/PK