Learn More
ABBERIOR INSTRUMENTS AMERICA LLC MOUNT SOLID ANTIFADE
Catalog No. NC1933856
Encompass_Preferred
abberior MOUNT SOLID ANTIFADE is an aqueous mounting medium designed for confocal and superresolution microscopy. The refractive index (before hardening) is identical to that of many biological samples (n=1.38). Specimen mounting using abberior MOUNT SOLID ANTIFADE is convenient since no dilution series is required. After specimen mounting, the embedding medium is hardening. So sealing of the mounted specimen with nail polish or similar is not required. abberior MOUNT SOLID ANTIFADE prevents photobleaching and preserves the signal of fluorescent labels. For scientific use only. Must not be used for diagnostic or medical purposes.
Catalog No. NC1933856 Supplier ABBERIOR INSTRUMENTS AMERICA LLC Supplier No. MM20132X15ML
abberior MOUNT SOLID ANTIFADE is an aqueous mounting medium designed for confocal and superresolution microscopy. The refractive index (before hardening) is identical to that of many biological samples (n=1.38). Specimen mounting using abberior MOUNT SOLID ANTIFADE is convenient since no dilution series is required. After specimen mounting, the embedding medium is hardening. So sealing of the mounted specimen with nail polish or similar is not required. abberior MOUNT SOLID ANTIFADE prevents photobleaching and preserves the signal of fluorescent labels. For scientific use only. Must not be used for diagnostic or medical purposes.