Learn More
Electron Microscopy Sciences SEM Mount Storage Box, Silicone Base
SDP
Catalog No. 5029564
Encompass_Preferred
A soft silicone base in a hinged plastic box. Base has 9 cavities 1" in diameter and 1/4"deep. Box holds 9 either 1" diameter or 1/8" diameter pin. Measurements: 4 3/4"(L)x3-5/8"(W)x 1/4"(H).
Use: 25 mm diameter or 3.2 mm pins
Catalog No. 50-295-64 Supplier Electron Microscopy Sciences Supplier No. 76525
A soft silicone base in a hinged plastic box. Base has 9 cavities 1" in diameter and 1/4"deep. Box holds 9 either 1" diameter or 1/8" diameter pin. Measurements: 4 3/4"(L)x3-5/8"(W)x 1/4"(H).
Use: 25 mm diameter or 3.2 mm pins