Learn More
Hampton Research HR4-313/Mounted CryoLoop - 20 micron, 0.025 - 0.05 mm - 25 pack
SDP
Catalog No. 500279
Encompass
HR4-313/Mounted CryoLoop - 20 micron, 0.025 - 0.05 mm - 25 pack. Please note items from this supplier are non-returnable.
Catalog No. 50-027-9 Supplier Hampton Research Supplier No. HR4313
HR4-313/Mounted CryoLoop - 20 micron, 0.025 - 0.05 mm - 25 pack. Please note items from this supplier are non-returnable.