Learn More
Hampton Research HR4-955/Mounted CryoLoop - 20 micron, 0.1 - 0.2 mm - 25 pack
SDP
Catalog No. 500299
Encompass
HR4-955/Mounted CryoLoop - 20 micron, 0.1 - 0.2 mm - 25 pack. Please note items from this supplier are non-returnable.
Catalog No. 50-029-9 Supplier Hampton Research Supplier No. HR4955
HR4-955/Mounted CryoLoop - 20 micron, 0.1 - 0.2 mm - 25 pack. Please note items from this supplier are non-returnable.