Learn More
Molecular Dimensions MOUNTED LITHOLOOPS 0.1MM
SDP
Catalog No. NC9611640
Encompass_Preferred
Protein Crystallography. 0.10mm LithoLoops mounted on 18mm stainless steel microtubes. Mounted in 18mm SPINE microtubes (pins). For use with Cryocaps. Just superglue the pin into a cyrocap and you're ready to go.
Catalog No. NC9611640 Supplier Molecular Dimensions Supplier No. MD7134
Protein Crystallography. 0.10mm LithoLoops mounted on 18mm stainless steel microtubes. Mounted in 18mm SPINE microtubes (pins). For use with Cryocaps. Just superglue the pin into a cyrocap and you're ready to go.