Learn More
Grainger Starrett Indicator Mounting Attachment, For 716X and 716MX Testers, 1 ea
Catalog No. 502787961
Encompass_Preferred
This indicator mounting attachment is designed to securely hold dovetail mount indicators, AGD indicators with 9.5 mm (3/8 in.) stems, and indicators utilizing a holder with a 4.7 mm (3/16 in.) shank. It serves as a replacement component for specific indicator testers and includes an integrated fine adjustment mechanism for precise zeroing.
  • Compatible with dovetail mount indicators
  • Accommodates AGD indicators with 9.5 mm (3/8 in.) stems
  • Supports indicators with 4.7 mm (3/16 in.) shank holders
  • Features fine adjustment for accurate zeroing
  • Designed as a replacement part for 716X and 716MX indicator testers
Catalog No. 50-278-7961 Supplier Grainger Supplier No. 197N95
This indicator mounting attachment is designed to securely hold dovetail mount indicators, AGD indicators with 9.5 mm (3/8 in.) stems, and indicators utilizing a holder with a 4.7 mm (3/16 in.) shank. It serves as a replacement component for specific indicator testers and includes an integrated fine adjustment mechanism for precise zeroing.
  • Compatible with dovetail mount indicators
  • Accommodates AGD indicators with 9.5 mm (3/8 in.) stems
  • Supports indicators with 4.7 mm (3/16 in.) shank holders
  • Features fine adjustment for accurate zeroing
  • Designed as a replacement part for 716X and 716MX indicator testers